Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAB4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GAB4 - GRB2 associated binding protein family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037814.1 | 2277 | UTR 3 | NP_001032903.1 | |||
XM_011546114.2 | 2277 | Intron | XP_011544416.1 | |||
XM_011546115.1 | 2277 | Intron | XP_011544417.1 | |||
XM_011546116.2 | 2277 | Intron | XP_011544418.1 | |||
XM_011546117.1 | 2277 | Intron | XP_011544419.1 | |||
XM_017028575.1 | 2277 | Intron | XP_016884064.1 | |||
XM_017028576.1 | 2277 | Intron | XP_016884065.1 |
Set Membership: |
HapMap |