Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300603 MIM: 314990 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
POF1B PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
POF1B - premature ovarian failure, 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307940.1 | 2197 | Intron | NP_001294869.1 | |||
NM_024921.3 | 2197 | UTR 3 | NP_079197.3 | |||
XM_005262203.3 | 2197 | UTR 3 | XP_005262260.1 |
ZNF711 - zinc finger protein 711 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |