Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604108 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAP7 - microtubule associated protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198608.1 | Intron | NP_001185537.1 | ||||
NM_001198609.1 | Intron | NP_001185538.1 | ||||
NM_001198611.1 | Intron | NP_001185540.1 | ||||
NM_001198614.1 | Intron | NP_001185543.1 | ||||
NM_001198615.1 | Intron | NP_001185544.1 | ||||
NM_001198616.1 | Intron | NP_001185545.1 | ||||
NM_001198617.1 | Intron | NP_001185546.1 | ||||
NM_001198618.1 | Intron | NP_001185547.1 | ||||
NM_001198619.1 | Intron | NP_001185548.1 | ||||
NM_003980.4 | Intron | NP_003971.1 | ||||
XM_006715598.2 | Intron | XP_006715661.1 | ||||
XM_006715599.2 | Intron | XP_006715662.1 | ||||
XM_006715600.2 | Intron | XP_006715663.1 | ||||
XM_006715601.3 | Intron | XP_006715664.1 | ||||
XM_011536243.2 | Intron | XP_011534545.1 | ||||
XM_011536244.2 | Intron | XP_011534546.1 | ||||
XM_011536245.1 | Intron | XP_011534547.1 | ||||
XM_011536246.2 | Intron | XP_011534548.1 | ||||
XM_017011471.1 | Intron | XP_016866960.1 |
Set Membership: |
HapMap |