Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGACCATGTTCTGTCTTCTCTGT[G/T]AGTTGAATCATCCTTTGGCCAGTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608143 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPAT4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GPAT4 - glycerol-3-phosphate acyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178819.3 | Intron | NP_848934.1 | ||||
XM_011544392.2 | Intron | XP_011542694.1 | ||||
XM_011544393.1 | Intron | XP_011542695.1 | ||||
XM_011544394.1 | Intron | XP_011542696.1 | ||||
XM_017013042.1 | Intron | XP_016868531.1 | ||||
XM_017013043.1 | Intron | XP_016868532.1 |
LOC102723729 - uncharacterized LOC102723729 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |