Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602548 MIM: 605071 MIM: 604784 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LKAAEAR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LKAAEAR1 - LKAAEAR motif containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6813 - microRNA 6813 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OPRL1 - opioid related nociceptin receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000913.5 | Intron | NP_000904.1 | ||||
NM_001200019.1 | Intron | NP_001186948.1 | ||||
NM_001318853.1 | Intron | NP_001305782.1 | ||||
NM_001318854.1 | Intron | NP_001305783.1 | ||||
NM_001318855.1 | Intron | NP_001305784.1 | ||||
NM_182647.3 | Intron | NP_872588.1 | ||||
XM_011528828.2 | Intron | XP_011527130.1 | ||||
XM_011528830.2 | Intron | XP_011527132.1 | ||||
XM_017027852.1 | Intron | XP_016883341.1 | ||||
XM_017027853.1 | Intron | XP_016883342.1 | ||||
XM_017027854.1 | Intron | XP_016883343.1 | ||||
XM_017027855.1 | Intron | XP_016883344.1 |
RGS19 - regulator of G-protein signaling 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TCEA2 - transcription elongation factor A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |