Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAGAGCGCCCTGGCGCCCTCCG[A/T]GGGCGTGCAGGCGGCGGGTGGCTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142983 | ||||||||||||||||||||
Literature Links: |
LINC01396 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LINC01396 - long intergenic non-protein coding RNA 1396 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MSX1 - msh homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002448.3 | 486 | Missense Mutation | GAG,GTG | E,V 84 | NP_002439.2 |