Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300562 | ||||||||||||||||||||
Literature Links: |
SLITRK4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLITRK4 - SLIT and NTRK like family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184749.2 | 3473 | UTR 3 | NP_001171678.1 | |||
NM_001184750.2 | 3473 | UTR 3 | NP_001171679.1 | |||
NM_173078.4 | 3473 | UTR 3 | NP_775101.1 | |||
XM_005262365.4 | 3473 | Intron | XP_005262422.1 | |||
XM_011531265.2 | 3473 | Intron | XP_011529567.1 |