Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 179520 | ||||||||||||||||||||
Literature Links: |
FAM212B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM212B - family with sequence similarity 212 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019099.4 | 5230 | UTR 3 | NP_061972.1 | |||
NM_198926.2 | 5230 | UTR 3 | NP_945120.1 | |||
XM_011541783.2 | 5230 | UTR 3 | XP_011540085.1 |
RAP1A - RAP1A, member of RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |