Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTTATCTCAGTTGCCATACTTTT[C/T]TTATTGCAGACCAGTTTGAGTTATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601191 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ERH - enhancer of rudimentary homolog (Drosophila) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC39A9 - solute carrier family 39 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252148.1 | Intron | NP_001239077.1 | ||||
NM_001252150.1 | Intron | NP_001239079.1 | ||||
NM_001252151.1 | Intron | NP_001239080.1 | ||||
NM_001252152.1 | Intron | NP_001239081.1 | ||||
NM_018375.4 | Intron | NP_060845.2 | ||||
XM_005267834.1 | Intron | XP_005267891.1 | ||||
XM_017021426.1 | Intron | XP_016876915.1 |
Set Membership: |
HapMap |