Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATTAATATTTATCAACTGAACC[C/G]AGTGGCAAGCACTCGGATCCCTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601963 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PWWP2A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PWWP2A - PWWP domain containing 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130864.1 | Intron | NP_001124336.1 | ||||
NM_001267035.1 | Intron | NP_001253964.1 | ||||
NM_052927.2 | Intron | NP_443159.1 | ||||
XM_005265811.3 | Intron | XP_005265868.1 | ||||
XM_005265812.2 | Intron | XP_005265869.1 | ||||
XM_006714817.3 | Intron | XP_006714880.1 | ||||
XM_011534424.2 | Intron | XP_011532726.1 | ||||
XM_017009009.1 | Intron | XP_016864498.1 | ||||
XM_017009010.1 | Intron | XP_016864499.1 | ||||
XM_017009011.1 | Intron | XP_016864500.1 | ||||
XM_017009012.1 | Intron | XP_016864501.1 |
TTC1 - tetratricopeptide repeat domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282500.1 | Intron | NP_001269429.1 | ||||
NM_003314.2 | Intron | NP_003305.1 |
Set Membership: |
HapMap |