Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGACGGACATGTTGCAAAGTCCTT[C/T]CCTCTTTCGGAGCTGAGTGAGAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR88 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
WDR88 - WD repeat domain 88 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173479.3 | Intron | NP_775750.3 | ||||
XM_011526449.2 | Intron | XP_011524751.1 | ||||
XM_011526450.2 | Intron | XP_011524752.1 | ||||
XM_017026282.1 | Intron | XP_016881771.1 | ||||
XM_017026283.1 | Intron | XP_016881772.1 |