Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAATAAAAACAGTCCCAACCTTCC[A/G]TCTAAATGTTGGTCCTCAGTTGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611732 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBOAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MBOAT1 - membrane bound O-acyltransferase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080480.2 | 4174 | UTR 3 | NP_001073949.1 | |||
XM_006714999.2 | 4174 | Intron | XP_006715062.1 | |||
XM_006715000.3 | 4174 | Intron | XP_006715063.1 | |||
XM_011514313.2 | 4174 | Intron | XP_011512615.1 |
Set Membership: |
HapMap |