Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604054 MIM: 604050 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACAA1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ACAA1 - acetyl-CoA acyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130410.1 | 1458 | Silent Mutation | GCA,GCG | A,A 324 | NP_001123882.1 | |
NM_001607.3 | 1458 | Silent Mutation | GCA,GCG | A,A 417 | NP_001598.1 | |
XM_006713122.1 | 1458 | Silent Mutation | GCA,GCG | A,A 357 | XP_006713185.1 | |
XM_006713123.2 | 1458 | Silent Mutation | GCA,GCG | A,A 265 | XP_006713186.1 | |
XM_011533650.2 | 1458 | Silent Mutation | GCA,GCG | A,A 272 | XP_011531952.1 |
DLEC1 - deleted in lung and esophageal cancer 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321153.1 | 1458 | Intron | NP_001308082.1 | |||
NM_007335.3 | 1458 | Intron | NP_031361.2 | |||
NM_007337.3 | 1458 | Intron | NP_031363.2 | |||
XM_006713438.3 | 1458 | Intron | XP_006713501.1 | |||
XM_006713439.3 | 1458 | Intron | XP_006713502.1 | |||
XM_006713440.3 | 1458 | Intron | XP_006713503.1 | |||
XM_011534317.2 | 1458 | Intron | XP_011532619.1 | |||
XM_011534318.2 | 1458 | Intron | XP_011532620.1 |
Set Membership: |
HapMap |