Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605317 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FOXP2 - forkhead box P2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172766.2 | Intron | NP_001166237.1 | ||||
NM_001172767.2 | Intron | NP_001166238.1 | ||||
NM_014491.3 | Intron | NP_055306.1 | ||||
NM_148898.3 | Intron | NP_683696.2 | ||||
NM_148899.3 | Intron | NP_683697.2 | ||||
NM_148900.3 | Intron | NP_683698.2 | ||||
XM_017012801.1 | Intron | XP_016868290.1 |