Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
43 submissions
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 601584 | ||||||||||||||||||||||||||
Literature Links: |
PTP4A2 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PTP4A2 - protein tyrosine phosphatase type IVA, member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195100.1 | Intron | NP_001182029.1 | ||||
NM_001195101.1 | Intron | NP_001182030.1 | ||||
NM_080391.3 | Intron | NP_536316.1 | ||||
XM_005271231.3 | Intron | XP_005271288.1 | ||||
XM_005271232.3 | Intron | XP_005271289.1 | ||||
XM_017002401.1 | Intron | XP_016857890.1 |
Set Membership: |
HapMap |