Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACACAGGGCCCTGCAATCAGCTC[C/T]GAGGCAGAACTGCCGCTGCGCTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605207 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP26B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CYP26B1 - cytochrome P450 family 26 subfamily B member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277742.1 | 2688 | UTR 3 | NP_001264671.1 | |||
NM_019885.3 | 2688 | UTR 3 | NP_063938.1 | |||
XM_005264433.4 | 2688 | UTR 3 | XP_005264490.1 | |||
XM_011532988.1 | 2688 | UTR 3 | XP_011531290.1 |
Set Membership: |
HapMap |