Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
39 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611368 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAEL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAEL - maelstrom spermatogenic transposon silencer | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286377.1 | Intron | NP_001273306.1 | ||||
NM_001286378.1 | Intron | NP_001273307.1 | ||||
NM_032858.2 | Intron | NP_116247.1 | ||||
XM_006711583.1 | Intron | XP_006711646.1 | ||||
XM_011510067.1 | Intron | XP_011508369.1 | ||||
XM_011510068.1 | Intron | XP_011508370.1 | ||||
XM_017002602.1 | Intron | XP_016858091.1 | ||||
XM_017002603.1 | Intron | XP_016858092.1 |