Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGGCAGCATCTACAACTAGATGC[A/C]AGTAGCAACCAAAATGTTAATGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615931 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRR16 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PRR16 - proline rich 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300783.1 | Intron | NP_001287712.1 | ||||
NM_001308087.1 | Intron | NP_001295016.1 | ||||
NM_001308088.1 | Intron | NP_001295017.1 | ||||
NM_016644.2 | Intron | NP_057728.1 | ||||
XM_011543452.2 | Intron | XP_011541754.1 | ||||
XM_011543453.2 | Intron | XP_011541755.1 | ||||
XM_011543454.2 | Intron | XP_011541756.1 |