Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCTTGATGGGATAACTATGCCT[C/T]GCAGATATGACTGGATGGAAAGAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
45 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604516 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD101 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CD101 - CD101 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256106.2 | Intron | NP_001243035.1 | ||||
NM_001256109.2 | Intron | NP_001243038.1 | ||||
NM_001256111.2 | Intron | NP_001243040.1 | ||||
NM_004258.5 | Intron | NP_004249.2 | ||||
XM_017002846.1 | Intron | XP_016858335.1 | ||||
XM_017002847.1 | Intron | XP_016858336.1 |
Set Membership: |
HapMap |