Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAN3 - PAN3 poly(A) specific ribonuclease subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175854.7 | Intron | NP_787050.6 | ||||
XM_005266332.3 | Intron | XP_005266389.1 | ||||
XM_005266333.3 | Intron | XP_005266390.1 | ||||
XM_005266334.1 | Intron | XP_005266391.1 | ||||
XM_011535032.2 | Intron | XP_011533334.1 | ||||
XM_011535033.2 | Intron | XP_011533335.1 | ||||
XM_011535034.2 | Intron | XP_011533336.1 | ||||
XM_017020528.1 | Intron | XP_016876017.1 | ||||
XM_017020529.1 | Intron | XP_016876018.1 | ||||
XM_017020530.1 | Intron | XP_016876019.1 | ||||
XM_017020531.1 | Intron | XP_016876020.1 | ||||
XM_017020532.1 | Intron | XP_016876021.1 | ||||
XM_017020533.1 | Intron | XP_016876022.1 |
PAN3-AS1 - PAN3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |