Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300267 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARHGEF6 - Rac/Cdc42 guanine nucleotide exchange factor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306177.1 | Intron | NP_001293106.1 | ||||
NM_004840.2 | Intron | NP_004831.1 | ||||
XM_005262499.2 | Intron | XP_005262556.1 | ||||
XM_011531412.2 | Intron | XP_011529714.1 | ||||
XM_011531413.1 | Intron | XP_011529715.1 | ||||
XM_011531414.1 | Intron | XP_011529716.1 | ||||
XM_011531415.2 | Intron | XP_011529717.1 | ||||
XM_011531416.2 | Intron | XP_011529718.1 | ||||
XM_011531417.1 | Intron | XP_011529719.1 | ||||
XM_017029975.1 | Intron | XP_016885464.1 | ||||
XM_017029976.1 | Intron | XP_016885465.1 |
Set Membership: |
HapMap |