Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGAAACGAACAGTCAGATGGAGA[G/T]GGAGGAGGGAAAGAAAAGGCTCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608386 MIM: 605208 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC155 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCDC155 - coiled-coil domain containing 155 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GFY - golgi associated olfactory signaling regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTH2 - parathyroid hormone 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178449.3 | Intron | NP_848544.1 |
SLC17A7 - solute carrier family 17 member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |