Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTGTTGTCTTTGAAAAGCTACA[A/C]ATGGAGCAGTGCAGAATACAGGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610133 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ST6GALNAC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ST6GALNAC3 - ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160011.1 | Intron | NP_001153483.1 | ||||
NM_152996.2 | Intron | NP_694541.2 | ||||
XM_006710553.3 | Intron | XP_006710616.1 | ||||
XM_017000937.1 | Intron | XP_016856426.1 | ||||
XM_017000938.1 | Intron | XP_016856427.1 | ||||
XM_017000939.1 | Intron | XP_016856428.1 | ||||
XM_017000940.1 | Intron | XP_016856429.1 | ||||
XM_017000941.1 | Intron | XP_016856430.1 | ||||
XM_017000942.1 | Intron | XP_016856431.1 |