Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTCTAGTGAGATGCTATATTTA[C/T]AAGTAAACATAAAAATCAGTACCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607247 MIM: 606635 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHODL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHODL - chondrolectin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMPRSS15 - transmembrane protease, serine 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002772.2 | Intron | NP_002763.2 | ||||
XM_011529654.1 | Intron | XP_011527956.1 | ||||
XM_011529655.1 | Intron | XP_011527957.1 | ||||
XM_011529656.2 | Intron | XP_011527958.1 | ||||
XM_011529657.1 | Intron | XP_011527959.1 | ||||
XM_011529658.1 | Intron | XP_011527960.1 | ||||
XM_011529659.1 | Intron | XP_011527961.1 |