Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGGCTGTTGTCCCGGGCTTGTC[A/C]CTCAAAGACGGGACAGATGTCTCCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609315 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102577426 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC102577426 - uncharacterized LOC102577426 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM7 - tripartite motif containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033342.3 | 2181 | Intron | NP_203128.1 | |||
NM_203293.2 | 2181 | UTR 3 | NP_976038.1 | |||
NM_203294.1 | 2181 | UTR 3 | NP_976039.1 | |||
NM_203295.1 | 2181 | UTR 3 | NP_976040.1 | |||
NM_203296.1 | 2181 | UTR 3 | NP_976041.1 | |||
NM_203297.1 | 2181 | UTR 3 | NP_976042.1 | |||
XM_017009903.1 | 2181 | UTR 3 | XP_016865392.1 | |||
XM_017009904.1 | 2181 | Intron | XP_016865393.1 |
Set Membership: |
HapMap |