Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAATCACTTTATCGGGAGTAGTT[G/T]TCTTCCTCACTACTGGGCAATGTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANP32AP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ANP32AP1 - acidic nuclear phosphoprotein 32 family member A pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DPH6 - diphthamine biosynthesis 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001141972.1 | Intron | NP_001135444.1 | ||||
NM_080650.3 | Intron | NP_542381.1 | ||||
XM_011522160.1 | Intron | XP_011520462.1 | ||||
XM_017022707.1 | Intron | XP_016878196.1 | ||||
XM_017022708.1 | Intron | XP_016878197.1 | ||||
XM_017022709.1 | Intron | XP_016878198.1 |