Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613437 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCHO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FCHO1 - FCH domain only 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161357.1 | Intron | NP_001154829.1 | ||||
NM_001161358.1 | Intron | NP_001154830.1 | ||||
NM_001161359.1 | Intron | NP_001154831.1 | ||||
NM_015122.2 | Intron | NP_055937.1 | ||||
XM_005259831.3 | Intron | XP_005259888.1 | ||||
XM_006722701.3 | Intron | XP_006722764.1 | ||||
XM_006722702.3 | Intron | XP_006722765.1 | ||||
XM_011527824.2 | Intron | XP_011526126.1 | ||||
XM_011527825.2 | Intron | XP_011526127.1 | ||||
XM_011527826.2 | Intron | XP_011526128.1 | ||||
XM_011527827.2 | Intron | XP_011526129.1 | ||||
XM_011527828.2 | Intron | XP_011526130.1 | ||||
XM_017026519.1 | Intron | XP_016882008.1 | ||||
XM_017026520.1 | Intron | XP_016882009.1 | ||||
XM_017026521.1 | Intron | XP_016882010.1 | ||||
XM_017026522.1 | Intron | XP_016882011.1 | ||||
XM_017026523.1 | Intron | XP_016882012.1 | ||||
XM_017026524.1 | Intron | XP_016882013.1 | ||||
XM_017026525.1 | Intron | XP_016882014.1 | ||||
XM_017026526.1 | Intron | XP_016882015.1 | ||||
XM_017026527.1 | Intron | XP_016882016.1 | ||||
XM_017026528.1 | Intron | XP_016882017.1 |