Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAGTCCTGGGTGGGCTCCTTTAC[A/G]AAAGCAGCAATTTATAGGGAGAGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604613 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBX18 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
TBX18 - T-box 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080508.2 | Intron | NP_001073977.1 | ||||
XM_006715602.3 | Intron | XP_006715665.2 | ||||
XM_011536247.2 | Intron | XP_011534549.2 | ||||
XM_011536248.2 | Intron | XP_011534550.2 | ||||
XM_011536249.2 | Intron | XP_011534551.2 | ||||
XM_017011472.1 | Intron | XP_016866961.1 | ||||
XM_017011473.1 | Intron | XP_016866962.1 |
Set Membership: |
HapMap |