Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAAACTGGAGCAGACAGGAGGTT[A/G]ATCACCTTTGCCTCCCTGAGGGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFCAB6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EFCAB6 - EF-hand calcium binding domain 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022785.3 | Intron | NP_073622.2 | ||||
NM_198856.2 | Intron | NP_942153.1 | ||||
XM_005261704.2 | Intron | XP_005261761.1 | ||||
XM_005261705.3 | Intron | XP_005261762.1 | ||||
XM_011530316.1 | Intron | XP_011528618.1 | ||||
XM_011530317.2 | Intron | XP_011528619.1 | ||||
XM_011530318.2 | Intron | XP_011528620.1 | ||||
XM_011530319.2 | Intron | XP_011528621.1 | ||||
XM_011530320.2 | Intron | XP_011528622.1 | ||||
XM_011530321.2 | Intron | XP_011528623.1 | ||||
XM_011530322.2 | Intron | XP_011528624.1 | ||||
XM_011530323.2 | Intron | XP_011528625.1 | ||||
XM_011530325.2 | Intron | XP_011528627.1 | ||||
XM_011530326.2 | Intron | XP_011528628.1 | ||||
XM_011530327.2 | Intron | XP_011528629.1 | ||||
XM_011530328.2 | Intron | XP_011528630.1 | ||||
XM_011530329.1 | Intron | XP_011528631.1 | ||||
XM_011530330.2 | Intron | XP_011528632.1 | ||||
XM_017028910.1 | Intron | XP_016884399.1 | ||||
XM_017028911.1 | Intron | XP_016884400.1 | ||||
XM_017028912.1 | Intron | XP_016884401.1 |
EFCAB6-AS1 - EFCAB6 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |