Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTTTTTGCTTCCACTTTAAGTTT[A/C]TTGATGCCAGCACATATCTTATTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142409 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HGF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HGF - hepatocyte growth factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000601.5 | 4218 | UTR 3 | NP_000592.3 | |||
NM_001010931.2 | 4218 | Intron | NP_001010931.1 | |||
NM_001010932.2 | 4218 | UTR 3 | NP_001010932.1 | |||
NM_001010933.2 | 4218 | Intron | NP_001010933.1 | |||
NM_001010934.2 | 4218 | Intron | NP_001010934.1 | |||
XM_006715956.2 | 4218 | UTR 3 | XP_006716019.1 | |||
XM_011516115.2 | 4218 | UTR 3 | XP_011514417.1 | |||
XM_017012097.1 | 4218 | Intron | XP_016867586.1 | |||
XM_017012098.1 | 4218 | Intron | XP_016867587.1 |
LOC100128317 - uncharacterized LOC100128317 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |