Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCCCCAAGCCAGGATGTCCCAA[A/G]CTTGGGTCCCCGGCCTCGCGCCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM92 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM92 - transmembrane protein 92 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168215.1 | 120 | Missense Mutation | ACT,GCT | T,A 4 | NP_001161687.1 | |
NM_153229.2 | 120 | Missense Mutation | ACT,GCT | T,A 4 | NP_694961.2 | |
XM_006721730.3 | 120 | Missense Mutation | ACT,GCT | T,A 4 | XP_006721793.1 |
TMEM92-AS1 - TMEM92 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |