Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601002 MIM: 609928 | ||||||||||||||||||||
Literature Links: |
GSS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GSS - glutathione synthetase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYH7B - myosin heavy chain 7B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020884.4 | Intron | NP_065935.3 | ||||
XM_006723840.3 | Intron | XP_006723903.1 | ||||
XM_011528941.2 | Intron | XP_011527243.1 | ||||
XM_011528947.2 | Intron | XP_011527249.1 | ||||
XM_011528948.2 | Intron | XP_011527250.1 | ||||
XM_017027986.1 | Intron | XP_016883475.1 | ||||
XM_017027987.1 | Intron | XP_016883476.1 |