Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGACACACAGGGGCAGCAAGTC[C/T]GAATTTTGCTCTCATCTCTGCTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605767 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PAG1 - phosphoprotein membrane anchor with glycosphingolipid microdomains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018440.3 | 9582 | UTR 3 | NP_060910.3 | |||
XM_017013640.1 | 9582 | UTR 3 | XP_016869129.1 | |||
XM_017013641.1 | 9582 | UTR 3 | XP_016869130.1 | |||
XM_017013642.1 | 9582 | UTR 3 | XP_016869131.1 | |||
XM_017013643.1 | 9582 | UTR 3 | XP_016869132.1 |