Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAAGAATAATGCACAAAAACTAT[C/T]ATAGCTCATCCATTCAAGTGGTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604912 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TAF2 - TATA-box binding protein associated factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003184.3 | Intron | NP_003175.1 | ||||
XM_006716621.3 | Intron | XP_006716684.1 | ||||
XM_011517259.2 | Intron | XP_011515561.1 | ||||
XM_017013779.1 | Intron | XP_016869268.1 | ||||
XM_017013780.1 | Intron | XP_016869269.1 | ||||
XM_017013781.1 | Intron | XP_016869270.1 | ||||
XM_017013782.1 | Intron | XP_016869271.1 | ||||
XM_017013783.1 | Intron | XP_016869272.1 | ||||
XM_017013784.1 | Intron | XP_016869273.1 | ||||
XM_017013785.1 | Intron | XP_016869274.1 | ||||
XM_017013786.1 | Intron | XP_016869275.1 | ||||
XM_017013787.1 | Intron | XP_016869276.1 | ||||
XM_017013788.1 | Intron | XP_016869277.1 | ||||
XM_017013789.1 | Intron | XP_016869278.1 | ||||
XM_017013790.1 | Intron | XP_016869279.1 | ||||
XM_017013791.1 | Intron | XP_016869280.1 |
Set Membership: |
HapMap |