Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATTTGCAGAATAAACATGGACT[C/T]AAACACTGAAACATGGAGTTTGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608235 MIM: 608489 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAL3ST4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GAL3ST4 - galactose-3-O-sulfotransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024637.4 | 2551 | Intron | NP_078913.3 |
GPC2 - glypican 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152742.2 | 2551 | UTR 3 | NP_689955.1 |
STAG3 - stromal antigen 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |