Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602715 | ||||||||||||||||||||
Literature Links: |
IPO9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IPO9 - importin 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LMOD1 - leiomodin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHISA4 - shisa family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198149.2 | 248 | Missense Mutation | AGG,ATG | R,M 64 | NP_937792.2 | |
XM_017000419.1 | 248 | Missense Mutation | AGG,ATG | R,M 64 | XP_016855908.1 |