Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCTGGCTCACAGTAGTTCAGTAG[A/C]TACTTGCTAAATAGTACTGAACTGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616091 MIM: 612166 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929718 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC101929718 - uncharacterized LOC101929718 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METTL17 - methyltransferase like 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029991.1 | Intron | NP_001025162.1 | ||||
NM_022734.2 | Intron | NP_073571.1 | ||||
XM_006720236.2 | Intron | XP_006720299.1 | ||||
XM_017021601.1 | Intron | XP_016877090.1 |
SLC39A2 - solute carrier family 39 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |