Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGAGCACTCCACCGTCATCGGCA[C/G]AGTCTGGCTCACCGTGCTTTTCATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600897 | ||||||||||||||||||||
Literature Links: |
GJA8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GJA8 - gap junction protein alpha 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005267.4 | 868 | Missense Mutation | ACA,AGA | T,R 23 | NP_005258.2 | |
XM_011509416.1 | 868 | Missense Mutation | ACA,AGA | T,R 23 | XP_011507718.1 | |
XM_011509417.1 | 868 | Missense Mutation | ACA,AGA | T,R 23 | XP_011507719.1 |