Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGCTTTTTCTGCTGTGTGATTTA[G/T]TCAAGAAATATTCAAATGTCAGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610677 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LSM14A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LSM14A - LSM14A, mRNA processing body assembly factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114093.1 | Intron | NP_001107565.1 | ||||
NM_015578.2 | Intron | NP_056393.2 | ||||
XM_005258719.4 | Intron | XP_005258776.1 | ||||
XM_005258720.4 | Intron | XP_005258777.1 | ||||
XM_005258721.4 | Intron | XP_005258778.1 | ||||
XM_011526708.2 | Intron | XP_011525010.1 | ||||
XM_017026575.1 | Intron | XP_016882064.1 | ||||
XM_017026576.1 | Intron | XP_016882065.1 | ||||
XM_017026577.1 | Intron | XP_016882066.1 |
Set Membership: |
HapMap |