Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCTCACACATGCCACAACCACA[C/G]ATGGTCCCCAGCAGGAACCCCAAGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606825 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNS3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TNS3 - tensin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022748.11 | 5485 | UTR 3 | NP_073585.8 | |||
XM_011515476.2 | 5485 | UTR 3 | XP_011513778.1 | |||
XM_011515477.2 | 5485 | UTR 3 | XP_011513779.1 | |||
XM_011515478.2 | 5485 | UTR 3 | XP_011513780.1 | |||
XM_011515479.2 | 5485 | UTR 3 | XP_011513781.1 | |||
XM_011515480.2 | 5485 | UTR 3 | XP_011513782.1 | |||
XM_011515481.2 | 5485 | UTR 3 | XP_011513783.1 | |||
XM_011515482.2 | 5485 | UTR 3 | XP_011513784.1 | |||
XM_011515483.2 | 5485 | UTR 3 | XP_011513785.1 | |||
XM_011515484.2 | 5485 | UTR 3 | XP_011513786.1 | |||
XM_017012537.1 | 5485 | UTR 3 | XP_016868026.1 | |||
XM_017012538.1 | 5485 | UTR 3 | XP_016868027.1 | |||
XM_017012539.1 | 5485 | UTR 3 | XP_016868028.1 |
Set Membership: |
HapMap |