Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATTTTCTTGGCAAAGATTTTTA[C/G]GCCATGTGAAAAGTATCTTTCAGGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600769 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSPAN8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
TSPAN8 - tetraspanin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004616.2 | Intron | NP_004607.1 | ||||
XM_006719583.3 | Intron | XP_006719646.1 | ||||
XM_017019913.1 | Intron | XP_016875402.1 |