Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTAGTTCTATGTGCTGCCCCCTT[C/T]CTGGAATGCTTTTCACACCCTCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604366 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNAI1 - dynein axonemal intermediate chain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281428.1 | Intron | NP_001268357.1 | ||||
NM_012144.3 | Intron | NP_036276.1 | ||||
XM_006716758.3 | Intron | XP_006716821.1 | ||||
XM_011517846.1 | Intron | XP_011516148.1 | ||||
XM_011517847.2 | Intron | XP_011516149.1 | ||||
XM_011517848.1 | Intron | XP_011516150.1 | ||||
XM_011517849.1 | Intron | XP_011516151.1 | ||||
XM_011517850.2 | Intron | XP_011516152.1 | ||||
XM_017014625.1 | Intron | XP_016870114.1 |
FAM219A - family with sequence similarity 219 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |