Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATATGGCACCTTGACATTGGAG[A/G]CTATAGCAGAAGCAGGAAGGTCATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608182 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNIP4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KCNIP4 - potassium voltage-gated channel interacting protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035003.1 | Intron | NP_001030175.1 | ||||
NM_001035004.1 | Intron | NP_001030176.1 | ||||
NM_025221.5 | Intron | NP_079497.2 | ||||
NM_147181.3 | Intron | NP_671710.1 | ||||
NM_147182.3 | Intron | NP_671711.1 | ||||
NM_147183.3 | Intron | NP_671712.1 | ||||
XM_005248190.4 | Intron | XP_005248247.1 | ||||
XM_011513882.2 | Intron | XP_011512184.1 | ||||
XM_011513883.2 | Intron | XP_011512185.1 | ||||
XM_011513884.2 | Intron | XP_011512186.1 | ||||
XM_011513885.2 | Intron | XP_011512187.1 | ||||
XM_011513886.2 | Intron | XP_011512188.1 | ||||
XM_011513887.2 | Intron | XP_011512189.1 | ||||
XM_011513889.2 | Intron | XP_011512191.1 | ||||
XM_017008653.1 | Intron | XP_016864142.1 |
KCNIP4-IT1 - KCNIP4 intronic transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |