Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCCTTAATCTGGTCAAACTTAA[A/G]TGCGATTGTGACCAGGCAGAGATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF536 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF536 - zinc finger protein 536 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014717.2 | Intron | NP_055532.1 | ||||
XM_011527554.2 | Intron | XP_011525856.1 | ||||
XM_011527555.2 | Intron | XP_011525857.1 | ||||
XM_011527557.2 | Intron | XP_011525859.1 | ||||
XM_011527558.2 | Intron | XP_011525860.1 | ||||
XM_011527560.2 | Intron | XP_011525862.1 | ||||
XM_017027527.1 | Intron | XP_016883016.1 | ||||
XM_017027528.1 | Intron | XP_016883017.1 | ||||
XM_017027529.1 | Intron | XP_016883018.1 | ||||
XM_017027530.1 | Intron | XP_016883019.1 | ||||
XM_017027531.1 | Intron | XP_016883020.1 | ||||
XM_017027532.1 | Intron | XP_016883021.1 | ||||
XM_017027533.1 | Intron | XP_016883022.1 | ||||
XM_017027534.1 | Intron | XP_016883023.1 | ||||
XM_017027535.1 | Intron | XP_016883024.1 | ||||
XM_017027536.1 | Intron | XP_016883025.1 | ||||
XM_017027537.1 | Intron | XP_016883026.1 | ||||
XM_017027538.1 | Intron | XP_016883027.1 | ||||
XM_017027539.1 | Intron | XP_016883028.1 | ||||
XM_017027540.1 | Intron | XP_016883029.1 | ||||
XM_017027541.1 | Intron | XP_016883030.1 | ||||
XM_017027542.1 | Intron | XP_016883031.1 | ||||
XM_017027543.1 | Intron | XP_016883032.1 |
Set Membership: |
HapMap |