Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGTTCCAACTAAGACTAAAGTG[A/G]CCGTTGATGAGAATAAAGCCAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611752 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf40 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C2orf40 - chromosome 2 open reading frame 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032411.2 | 199 | Missense Mutation | ACC,GCC | T,A 52 | NP_115787.1 | |
XM_006712799.3 | 199 | Missense Mutation | ACC,GCC | T,A 16 | XP_006712862.1 | |
XM_011511993.2 | 199 | Missense Mutation | ACC,GCC | T,A 16 | XP_011510295.1 | |
XM_017005106.1 | 199 | Missense Mutation | ACC,GCC | T,A 54 | XP_016860595.1 |