Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603444 | ||||||||||||||||||||
Literature Links: |
FUBP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FUBP1 - far upstream element binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303433.1 | Intron | NP_001290362.1 | ||||
NM_003902.4 | Intron | NP_003893.2 | ||||
XM_011542391.2 | Intron | XP_011540693.1 | ||||
XM_011542392.2 | Intron | XP_011540694.1 | ||||
XM_011542393.2 | Intron | XP_011540695.1 | ||||
XM_017002739.1 | Intron | XP_016858228.1 | ||||
XM_017002740.1 | Intron | XP_016858229.1 | ||||
XM_017002741.1 | Intron | XP_016858230.1 | ||||
XM_017002742.1 | Intron | XP_016858231.1 | ||||
XM_017002743.1 | Intron | XP_016858232.1 | ||||
XM_017002744.1 | Intron | XP_016858233.1 |