Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMG4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PSMG4 - proteasome assembly chaperone 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128591.1 | Intron | NP_001122063.1 | ||||
NM_001128592.1 | Intron | NP_001122064.1 | ||||
NM_001135750.1 | Intron | NP_001129222.1 | ||||
XM_011514591.2 | Intron | XP_011512893.1 | ||||
XM_011514593.2 | Intron | XP_011512895.1 | ||||
XM_011514594.2 | Intron | XP_011512896.1 | ||||
XM_011514595.2 | Intron | XP_011512897.1 |
Set Membership: |
HapMap |