Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCATCCCGTGGGCACAGGTCATA[C/T]CTGCCTCGAGCAGTTGGGCACTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300846 MIM: 300171 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD99L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CD99L2 - CD99 molecule like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184808.1 | Intron | NP_001171737.1 | ||||
NM_001242614.1 | Intron | NP_001229543.1 | ||||
NM_031462.3 | Intron | NP_113650.2 | ||||
NM_134445.3 | Intron | NP_604394.1 | ||||
NM_134446.3 | Intron | NP_604395.1 | ||||
XM_011531202.2 | Intron | XP_011529504.1 | ||||
XM_017029890.1 | Intron | XP_016885379.1 | ||||
XM_017029891.1 | Intron | XP_016885380.1 |
MTMR1 - myotubularin related protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |