Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTACAAGCTTTGATTTGGGTGAA[A/T]GTCGTGGGAAGAAGCAATATTGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC61 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC61 - coiled-coil domain containing 61 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267723.1 | 264 | Intron | NP_001254652.1 | |||
XM_005259191.3 | 264 | Intron | XP_005259248.1 | |||
XM_005259192.4 | 264 | UTR 5 | XP_005259249.1 | |||
XM_011527257.2 | 264 | Intron | XP_011525559.1 |