Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGATTGAAGAAAATGAATAAAAT[A/C]GGAAGGATATGTGAGATAATACCAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
ISY1-RAB43 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ISY1-RAB43 - ISY1-RAB43 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204890.1 | Intron | NP_001191819.1 |
RAB43 - RAB43, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204883.1 | Intron | NP_001191812.1 | ||||
NM_001204884.1 | Intron | NP_001191813.1 | ||||
NM_001204885.1 | Intron | NP_001191814.1 | ||||
NM_001204886.1 | Intron | NP_001191815.1 | ||||
NM_001204887.1 | Intron | NP_001191816.1 | ||||
NM_001204888.1 | Intron | NP_001191817.1 | ||||
NM_198490.2 | Intron | NP_940892.1 |